2-(1-Pyrrolidinyl)-3-pyridinamine 95%
Lieferant: Apollo Scientific
OR913825-1G
APOSOR913825-1GEA
364
EUR
APOSOR913825-1G
2-(1-Pyrrolidinyl)-3-pyridinamine 95%
2-(1-Pyrrolidinyl)-3-pyridinamine
Formel:
C₉H₁₃N₃ Molecular Weight: 163,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08700176 CAS-Nummer: 5028-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...