(2S)-1-(1-Methylethyl)azetidine-2-carboxylic acid

Lieferant: Apollo Scientific

OR913830-1G
APOSOR913830-1GEA 811 EUR
APOSOR913830-1G
(2S)-1-(1-Methylethyl)azetidine-2-carboxylic acid
(2S)-1-(1-Methylethyl)azetidine-2-carboxylic acid
Formel: C₇H₁₃NO₂
Molecular Weight: 143,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11053659
CAS-Nummer: 255882-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR