(S)-3-Amino-3-(3-nitrophenyl)propionic acid 97%

Lieferant: Apollo Scientific
Achtung

OR913926-1G OR913926-5G
APOSOR913926-1GEA 178 EUR
APOSOR913926-1G APOSOR913926-5G
(S)-3-Amino-3-(3-nitrophenyl)propionic acid 97%
(S)-3-Amino-3-(3-nitrophenyl)propionic acid
Formel: C₉H₁₀N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD04113693
CAS-Nummer: 734529-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR