3-(Ethylamino)-4-nitrophenol 97%

Lieferant: Apollo Scientific

OR913948-10G OR913948-1G OR913948-5G
APOSOR913948-10GEA 754 EUR
APOSOR913948-10G APOSOR913948-1G APOSOR913948-5G
3-(Ethylamino)-4-nitrophenol 97%
3-(Ethylamino)-4-nitrophenol
Formel: C₈H₁₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD25121745
CAS-Nummer: 25186-40-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR