(S)-Amino-cyclopentyl-acetic acid methyl ester hydrochloride 97%

Lieferant: Apollo Scientific

Synonyme: (S)-Methyl 2-amino-2-cyclopentylacetate hydrochloride

OR913988-1G
APOSOR913988-1GEA 60.9 EUR
APOSOR913988-1G
(S)-Amino-cyclopentyl-acetic acid methyl ester hydrochloride 97%
(S)-Amino-cyclopentyl-acetic acid methyl ester hydrochloride
Formel: C₈H₁₅NO₂·ClH
Molecular Weight: 193,671 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800494
CAS-Nummer: 14328-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR