(S)-4-Boc-2-(aminomethyl)morpholine 97%

Lieferant: Apollo Scientific

Synonyme: (S)-tert-Butyl 2-(aminomethyl)morpholine-4-carboxylate

OR914006-1G OR914006-250MG
APOSOR914006-1GEA 97.3 EUR
APOSOR914006-1G APOSOR914006-250MG
(S)-4-Boc-2-(aminomethyl)morpholine 97%
(S)-4-Boc-2-(aminomethyl)morpholine
Formel: C₁₀H₂₀N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11707017
CAS-Nummer: 879403-42-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR