1-(2-Chloroethyl)imidazolidin-2-one 95%
Lieferant: Apollo Scientific
OR914077-1G
APOSOR914077-1GEA
128
EUR
APOSOR914077-1G
1-(2-Chloroethyl)imidazolidin-2-one 95%
1-(2-Chloroethyl)imidazolidin-2-one
Formel:
C₅H₉ClN₂O Molecular Weight: 148,59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09029607 CAS-Nummer: 2387-20-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...