4-Imidazol-1-ylmethylphenylamine 95%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-[(1H-Imidazol-1-yl)methyl]aniline

OR9141-5G
APOSOR9141-5GEA 163 EUR
APOSOR9141-5G
4-Imidazol-1-ylmethylphenylamine 95%
4-Imidazol-1-ylmethylphenylamine
Formel: C₁₀H₁₁N₃
Molecular Weight: 173,2144 g/mol
Schmelzpunkt: 128…130 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06660785
CAS-Nummer: 56643-85-7
UN: 2923
ADR: 8+6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR