1-(4-Methylphenyl)-1H-pyrrole 95%

Lieferant: Apollo Scientific

OR914123-1G OR914123-5G
APOSOR914123-1GEA 219 EUR
APOSOR914123-1G APOSOR914123-5G
1-(4-Methylphenyl)-1H-pyrrole 95%
1-(4-Methylphenyl)-1H-pyrrole
Formel: C₁₁H₁₁N
Lagertemperatur: Raumtemperatur
MDL: MFCD00799565
CAS-Nummer: 827-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR