1,2,3,4-Tetrahydropyrrolo[1,2-a]pyrazine 95%

Lieferant: Apollo Scientific

OR914166-1G
APOSOR914166-1GEA 548 EUR
APOSOR914166-1G
1,2,3,4-Tetrahydropyrrolo[1,2-a]pyrazine 95%
1,2,3,4-Tetrahydropyrrolo[1,2-a]pyrazine
Formel: C₇H₁₀N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00296976
CAS-Nummer: 71257-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR