2-Amino-4,5-dimethylthiophene-3-carbonitrile 98%

Lieferant: Apollo Scientific

OR914214-1G
APOSOR914214-1GEA 63.8 EUR
APOSOR914214-1G
2-Amino-4,5-dimethylthiophene-3-carbonitrile 98%
2-Amino-4,5-dimethylthiophene-3-carbonitrile
Formel: C₇H₈N₂S
Molecular Weight: 152.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00720189
CAS-Nummer: 4651-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR