3-Methoxy-2-nitrophenol 98%

Lieferant: Apollo Scientific
Achtung

OR914259-1G OR914259-250MG OR914259-5G
APOSOR914259-1GEA 284 EUR
APOSOR914259-1G APOSOR914259-250MG APOSOR914259-5G
3-Methoxy-2-nitrophenol 98%
3-Methoxy-2-nitrophenol
Formel: C₇H₇NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD16876801
CAS-Nummer: 3114-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR