(R)-4-Bromostyrene oxide 95%

Lieferant: Apollo Scientific

OR914304-1G
APOSOR914304-1GEA 609 EUR
APOSOR914304-1G
(R)-4-Bromostyrene oxide 95%
(R)-4-Bromostyrene oxide
Formel: C₈H₇BrO
Molecular Weight: 199.045 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08277298
CAS-Nummer: 62566-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR