3-Bromo-5-methylpyrazin-2-amine 95%
Lieferant: Apollo Scientific
OR914309-1G
APOSOR914309-1GEA
0
EUR
APOSOR914309-1G
3-Bromo-5-methylpyrazin-2-amine 95%
3-Bromo-5-methylpyrazin-2-amine
Formel:
C5H6BrN3 Lagertemperatur: Raumtemperatur |
MDL:
MFCD09750037 CAS-Nummer: 74290-65-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...