(R)-5-Bromomethyl-2-pyrrolidinone 95%

Lieferant: Apollo Scientific

Synonyme: (R)-5-(Bromomethyl)pyrrolidin-2-one

OR914328-5G
APOSOR914328-5GEA 242 EUR
APOSOR914328-5G
(R)-5-Bromomethyl-2-pyrrolidinone 95%
(R)-5-Bromomethyl-2-pyrrolidinone
Formel: C₅H₈BrNO
Molecular Weight: 178.027 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06799465
CAS-Nummer: 98612-60-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR