4-Pentylcyclohexanon 98%

Lieferant: Apollo Scientific

OR914341-100G
APOSOR914341-100GEA 120 EUR
APOSOR914341-100G
4-Pentylcyclohexanon 98%
4-Pentylcyclohexanone
Formel: C₁₁H₂₀O
Molecular Weight: 168.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00059581
CAS-Nummer: 61203-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR