Dimethyl-2,6-pyridindicarboxylat 97%

Lieferant: Apollo Scientific

Synonyme: Dimethyl pyridine-2,6-dicarboxylate

OR914344-100G
APOSOR914344-100GEA 89.9 EUR
APOSOR914344-100G
Dimethyl-2,6-pyridindicarboxylat 97%
Dimethyl-2,6-pyridindicarboxylat
Formel: C₉H₉NO₄
Molecular Weight: 195.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00134493
CAS-Nummer: 5453-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR