5-nitronaphthalen-1-amine 95%

Lieferant: Apollo Scientific

OR914366-1G
APOSOR914366-1GEA 632 EUR
APOSOR914366-1G
5-nitronaphthalen-1-amine 95%
5-nitronaphthalen-1-amine
Formel: C₁₀H₈N₂O₂
Molecular Weight: 188,183 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00230525
CAS-Nummer: 3272-91-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR