4-Cyanophenylacetic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-(4-Cyanophenyl)acetic acid

OR914390-25G
APOSOR914390-25GEA 143 EUR
APOSOR914390-25G
4-Cyanophenylacetic acid 98%
4-Cyanophenylacetic acid
Formel: C₉H₇NO₂
Molecular Weight: 161.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06798066
CAS-Nummer: 5462-71-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR