2-Hydroxy-2-(4-methylphenyl)acetic acid 98%

Lieferant: Apollo Scientific

OR914401-1G OR914401-25G OR914401-5G
APOSOR914401-1GEA 0 EUR
APOSOR914401-1G APOSOR914401-25G APOSOR914401-5G
2-Hydroxy-2-(4-methylphenyl)acetic acid 98%
2-Hydroxy-2-(4-methylphenyl)acetic acid
Formel: C₉H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00961860
CAS-Nummer: 31284-89-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR