3-Benzoylbenzyl bromide

Lieferant: Apollo Scientific
Achtung

OR914410-25G OR914410-5G
APOSOR914410-25GEA 304 EUR
APOSOR914410-25G APOSOR914410-5G
3-Benzoylbenzyl bromide
3-Benzoylbenzyl bromide
Formel: C₁₄H₁₁BrO
Lagertemperatur: Raumtemperatur
MDL: MFCD00044727
CAS-Nummer: 22071-24-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR