(Benzo[b]furan-3-yl)acetic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Carboxymethyl)benzo[b]furan, 2-(Benzofuran-3-yl)acetic acid, (1-Benzofuran-3-yl)acetic acid

OR9145-250MG
APOSOR9145-250MGEA 31.9 EUR
APOSOR9145-250MG
(Benzo[b]furan-3-yl)acetic acid 97%
(Benzo[b]furan-3-yl)acetic acid
Formel: C₁₀H₈O₃
Molecular Weight: 176,169 g/mol
Schmelzpunkt: 90…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04114279
CAS-Nummer: 64175-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR