2,6-Dichloranisol 98%

Lieferant: Apollo Scientific

Synonyme: 1,3-Dichloro-2-methoxybenzene

OR914523-100G
APOSOR914523-100GEA 142 EUR
APOSOR914523-100G
2,6-Dichloranisol 98%
2,6-Dichloranisol
Formel: C₇H₆Cl₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00000575
CAS-Nummer: 1984-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR