4'-(Dimethylamino)acetophenon 98%

Lieferant: Apollo Scientific

OR914527-100G OR914527-25G
APOSOR914527-100GEA 43 EUR
APOSOR914527-100G APOSOR914527-25G
4'-(Dimethylamino)acetophenon 98%
4'-(Dimethylamino)acetophenon
Formel: C₁₀H₁₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00017246
CAS-Nummer: 2124-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR