3-(Hydroxymethyl)-2-hydroxynaphthalin 98%

Lieferant: Apollo Scientific

Synonyme: 3-(Hydroxymethyl)naphthalen-2-ol

OR914542-1G
APOSOR914542-1GEA 528 EUR
APOSOR914542-1G
3-(Hydroxymethyl)-2-hydroxynaphthalin 98%
3-(Hydroxymethyl)-2-hydroxynaphthalin
Formel: C₁₁H₁₀O₂
Molecular Weight: 174.195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11504950
CAS-Nummer: 30159-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR