3-[1,2,4]Triazol-1-ylmethylbenzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR9147-1G
APOSOR9147-1GEA
139
EUR
APOSOR9147-1G
3-[1,2,4]Triazol-1-ylmethylbenzoic acid 97%
3-[1,2,4]Triazol-1-ylmethylbenzoic acid
Formel:
C₁₀H₉N₃O₂ Molecular Weight: 203,2 g/mol Schmelzpunkt: 209…210 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07186456 CAS-Nummer: 857284-23-2 UN: 1325 ADR: 4.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...