3-(Fur-2-yl)-1-methyl-1H-pyrazole-5-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(5-Carboxy-1-methyl-1H-pyrazol-3-yl)furan, 5-Carboxy-3-(fur-2-yl)-1-methyl-1H-pyrazole

OR9148-1G
APOSOR9148-1GEA 395 EUR
APOSOR9148-1G
3-(Fur-2-yl)-1-methyl-1H-pyrazole-5-carboxylic acid 97%
3-(Fur-2-yl)-1-methyl-1H-pyrazole-5-carboxylic acid
Formel: C₉H₈N₂O₃
Molecular Weight: 192.171 g/mol
Schmelzpunkt: 130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08271950
CAS-Nummer: 859851-00-6
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR