1-(Triisopropylsilyl)pyrrole-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR915073-1G OR915073-5G
APOSOR915073-1GEA 241 EUR
APOSOR915073-1G APOSOR915073-5G
1-(Triisopropylsilyl)pyrrole-3-carbaldehyde
1-(Triisopropylsilyl)pyrrole-3-carbaldehyde
Formel: C₁₄H₂₅NOSi
Lagertemperatur: Raumtemperatur
MDL: MFCD28515390
CAS-Nummer: 90971-76-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR