2-Phenoxybenzoyl chloride 90%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9152-1G
OR9152-250MG
APOSOR9152-1GEA
306
EUR
APOSOR9152-1G
APOSOR9152-250MG
2-Phenoxybenzoyl chloride 90%
2-Phenoxybenzoyl chloride
Formel:
C₁₃H₉ClO₂ Schmelzpunkt: 37…39 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03424711 CAS-Nummer: 40501-36-8 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...