5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-inden-2-ol 98%

Lieferant: Apollo Scientific
Achtung

OR915673-1G
APOSOR915673-1GEA 1540 EUR
APOSOR915673-1G
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-inden-2-ol 98%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-inden-2-ol
Formel: C₁₅H₂₁BO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD27936516
CAS-Nummer: 1857348-77-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR