1-Methyl-5-phenyl-1H-pyrazol-3-carbaldehyd 97%

Lieferant: Apollo Scientific
Achtung

OR9158-250MG
APOSOR9158-250MGEA 154 EUR
APOSOR9158-250MG
1-Methyl-5-phenyl-1H-pyrazol-3-carbaldehyd 97%
1-Methyl-5-phenyl-1H-pyrazol-3-carbaldehyd
Formel: C₁₁H₁₀N₂O
Schmelzpunkt: 106…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08271935
CAS-Nummer: 124344-94-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR