2-Hydroxy-2,4-dimethylpentanoic acid 95%

Lieferant: Apollo Scientific

OR916220-1G
APOSOR916220-1GEA 609 EUR
APOSOR916220-1G
2-Hydroxy-2,4-dimethylpentanoic acid 95%
2-Hydroxy-2,4-dimethylpentanoic acid
Formel: C₇H₁₄O₃
Molecular Weight: 146,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14601961
CAS-Nummer: 3639-23-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR