N-(3-Chloropropyl)dibutylamine 95%

Lieferant: Apollo Scientific

OR916239-100G
APOSOR916239-100GEA 109 EUR
APOSOR916239-100G
N-(3-Chloropropyl)dibutylamine 95%
N-(3-Chloropropyl)dibutylamine
Formel: C₁₁H₂₄ClN
Lagertemperatur: Kühlschrank
MDL: MFCD13185966
CAS-Nummer: 36421-15-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR