Ethyl 4,5-dimethylisoxazole-3-carboxylate 95%

Lieferant: Apollo Scientific

OR916246-1G
APOSOR916246-1GEA 464 EUR
APOSOR916246-1G
Ethyl 4,5-dimethylisoxazole-3-carboxylate 95%
Ethyl 4,5-dimethylisoxazole-3-carboxylate
Formel: C₈H₁₁NO₃
Molecular Weight: 169,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18836299
CAS-Nummer: 160850-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR