(R)-2-Methylbutane-2-sulphinamide 95%

Lieferant: Apollo Scientific

OR916267-1G
APOSOR916267-1GEA 400 EUR
APOSOR916267-1G
(R)-2-Methylbutane-2-sulphinamide 95%
(R)-2-Methylbutane-2-sulphinamide
Formel: C₅H₁₃NOS
Molecular Weight: 135.228 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11707200
CAS-Nummer: 446021-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR