3-Methyl-2-hexanon 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 1-methylbutyl ketone, 3-Methylhexan-2-one

OR916391-1G
APOSOR916391-1GEA 394 EUR
APOSOR916391-1G
3-Methyl-2-hexanon 95%
3-Methyl-2-hexanon
Formel: C₇H₁₄O
Molecular Weight: 114.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039948
CAS-Nummer: 2550-21-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR