1-Methyl-1H-indole-5-sulphonyl chloride 90+%
Lieferant: Apollo Scientific
|
Gefahr
|
OR9164-1G
APOSOR9164-1GEA
308
EUR
APOSOR9164-1G
1-Methyl-1H-indole-5-sulphonyl chloride 90+%
1-Methyl-1H-indole-5-sulphonyl chloride
Formel:
C₉H₈ClNO₂S Schmelzpunkt: 79…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08271907 CAS-Nummer: 859850-75-2 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...