16-Hentriacontanon 95%

Lieferant: Apollo Scientific

OR916407-25G
APOSOR916407-25GEA 567 EUR
APOSOR916407-25G
16-Hentriacontanon 95%
16-Hentriacontanon
Formel: C₃₁H₆₂O
Schmelzpunkt: 84 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00059222
CAS-Nummer: 502-73-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR