7-Chloro-D-tryptophan 97%

Lieferant: Apollo Scientific

OR916429-250MG
APOSOR916429-250MGEA 0 EUR
APOSOR916429-250MG
7-Chloro-D-tryptophan 97%
7-Chloro-D-tryptophan
Formel: C₁₁H₁₁ClN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09264323
CAS-Nummer: 75102-74-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR