6-Ethyl-2-naphthalenol 95%

Lieferant: Apollo Scientific

OR916439-1G
APOSOR916439-1GEA 194 EUR
APOSOR916439-1G
6-Ethyl-2-naphthalenol 95%
6-Ethyl-2-naphthalenol
Formel: C₁₂H₁₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD07368645
CAS-Nummer: 1999-64-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR