4-Hydroxyindoline 98%
Lieferant: Apollo Scientific
Synonyme:
Indolin-4-ol
OR916446-1G
APOSOR916446-1GEA
249
EUR
APOSOR916446-1G
4-Hydroxyindoline 98%
4-Hydroxyindoline
Formel:
C₈H₉NO Molecular Weight: 135,163 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09056797 CAS-Nummer: 85926-99-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...