(3,5-Dichloropyridin-4-yl)methanamine 95%

Lieferant: Apollo Scientific

OR916476-1G
APOSOR916476-1GEA 171 EUR
APOSOR916476-1G
(3,5-Dichloropyridin-4-yl)methanamine 95%
(3,5-Dichloropyridin-4-yl)methanamine
Formel: C₆H₆Cl₂N₂
Molecular Weight: 177,031 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18254248
CAS-Nummer: 418795-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR