3-Bromo-5-hydroxybenzaldehyde

Lieferant: Apollo Scientific
Achtung

OR916487-5G
APOSOR916487-5GEA 220 EUR
APOSOR916487-5G
3-Bromo-5-hydroxybenzaldehyde
3-Bromo-5-hydroxybenzaldehyde
Formel: C₇H₅BrO₂
Molecular Weight: 201.017 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797981
CAS-Nummer: 199177-26-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR