tert-Butylbenzoat 95%

Lieferant: Apollo Scientific

Synonyme: Benzoic acid tert-butyl ester

OR916491-100G
APOSOR916491-100GEA 161 EUR
APOSOR916491-100G
tert-Butylbenzoat 95%
tert-Butylbenzoat
Formel: C₁₁H₁₄O₂
Molecular Weight: 178.2277 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00048244
CAS-Nummer: 774-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR