1-Benzylpiperidine 98%

Lieferant: Apollo Scientific

OR916524-100G
APOSOR916524-100GEA 186 EUR
APOSOR916524-100G
1-Benzylpiperidine 98%
1-Benzylpiperidine
Formel: C₁₂H₁₇N
Molecular Weight: 175.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00224901
CAS-Nummer: 2905-56-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR