4-Amylcyclohexylamine (cis- and trans- mixture) 95%

Lieferant: Apollo Scientific

OR916545-5G
APOSOR916545-5GEA 632 EUR
APOSOR916545-5G
4-Amylcyclohexylamine (cis- and trans- mixture) 95%
4-Amylcyclohexylamine (cis- and trans- mixture)
Formel: C₁₁H₂₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD07778407
CAS-Nummer: 38793-01-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR