Methyl 4-methoxy-2,5-dimethylbenzoate 95%
Lieferant: Apollo Scientific
OR916559-1G
APOSOR916559-1GEA
487
EUR
APOSOR916559-1G
Methyl 4-methoxy-2,5-dimethylbenzoate 95%
Methyl 4-methoxy-2,5-dimethylbenzoate
Formel:
C₁₁H₁₄O₃ Molecular Weight: 194,227 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06203806 CAS-Nummer: 58106-28-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...