(R)-3-Aminopyrrolidin-2-one hydrochloride ≥97%

Lieferant: Apollo Scientific

Synonyme: (R)-3-Amino-2-pyrrolidone Hydrochloride

OR916588-250MG
APOSOR916588-250MGEA 132 EUR
APOSOR916588-250MG
(R)-3-Aminopyrrolidin-2-one hydrochloride ≥97%
(R)-3-Aminopyrrolidin-2-one hydrochloride
Formel: C₄H₈N₂O·ClH
Molecular Weight: 136,58 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD14635509
CAS-Nummer: 223407-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR