3-Chloro-4-(hydroxymethyl)phenol 95%

Lieferant: Apollo Scientific

OR916640-1G
APOSOR916640-1GEA 522 EUR
APOSOR916640-1G
3-Chloro-4-(hydroxymethyl)phenol 95%
3-Chloro-4-(hydroxymethyl)phenol
Formel: C₇H₇ClO₂
Molecular Weight: 158,582 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00238558
CAS-Nummer: 171569-42-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR