6-Methoxy-1-naphthol 95%

Lieferant: Apollo Scientific

Synonyme: 6-Methoxynaphthalen-1-ol

OR916643-5G
APOSOR916643-5GEA 200 EUR
APOSOR916643-5G
6-Methoxy-1-naphthol 95%
6-Methoxy-1-naphthol
Formel: C₁₁H₁₀O₂
Molecular Weight: 174,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03695468
CAS-Nummer: 22604-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR